Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2277

Sigma-Aldrich

A192621

≥98 (HPLC)

Synonym(s):

(2R,3R,4S)-4-(1,3-Benzodioxol-5-yl)-1-[2-[(2,6-diethylphenyl)amino]-2-oxoethyl]-2-(4-propoxyphenyl)-3-pyrrolidinecarboxylic acid, A-192621

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H38N2O6
CAS Number:
Molecular Weight:
558.66
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(NC1=C(CC)C=CC=C1CC)CN(C[C@H](C2=CC3=C(OCO3)C=C2)[C@H]4C(O)=O)[C@H]4C5=CC=C(OCCC)C=C5

InChI

1S/C33H38N2O6/c1-4-16-39-25-13-10-23(11-14-25)32-30(33(37)38)26(24-12-15-27-28(17-24)41-20-40-27)18-35(32)19-29(36)34-31-21(5-2)8-7-9-22(31)6-3/h7-15,17,26,30,32H,4-6,16,18-20H2,1-3H3,(H,34,36)(H,37,38)/t26-,30-,32+/m1/s1

InChI key

LQEHCKYYIXQEBM-FUKIBTTHSA-N

Biochem/physiol Actions

A192621 is a selective; orally available ETB endothelin receptor antagonist. A192621 has been shown to cause an increase in arterial blood pressure and to promote apoptosis in human pulmonary arterial smooth muscle cells.
Selective; orally available ETB endothelin receptor antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerry L Wessale et al.
Clinical science (London, England : 1979), 103 Suppl 48, 112S-117S (2002-08-24)
Endothelins (ETs), 21-amino-acid peptides involved in the pathogenesis of various diseases, bind to ET(A) and ET(B) receptors to initiate their effects. Based on the same core structure, we have developed four small-molecule ET receptor antagonists, ABT-627 (atrasentan), ABT-546, A-182086 and
Eileen Miller et al.
Hypertension (Dallas, Tex. : 1979), 69(2), 275-285 (2016-12-29)
The role of smooth muscle endothelinB (ETB) receptors in regulating vascular function, blood pressure (BP), and neointimal remodeling has not been established. Selective knockout mice were generated to address the hypothesis that loss of smooth muscle ETB receptors would reduce
Satoshi Sakai et al.
Life sciences, 159, 116-120 (2016-03-30)
Vascular remodeling results from aberrations in the balance between cell proliferation and death, which is seen in the obstructive vasculature of pulmonary arterial hypertension (PAH). Endothelin (ET)-1 has a potent proliferative activity on vascular smooth muscle cells, and ET receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service