Skip to Content
MilliporeSigma

SML2124

Ned-K

≥98% (HPLC)

Synonym(s):

(1R,3S)-1-(3-((4-(2-Cyanophenyl)piperazin-1-yl)methyl)-4-methoxyphenyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C31H31N5O3
CAS Number:
Molecular Weight:
521.61
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

Ned-K is a specific and potent inhibitor of NAADP (nicotinic acid adenine dinucleotide phosphate) signaling that suppresses Ca2+ oscillations via two-pore channels (TPC1) and opening of the mitochondrial permeability transition pore (mPTP) in cardiomyocytes during simulated ischaemia and reperfusion. Ned-K protected cardiomyocytes from cell death during ischaemia and reperfusion injury in rats.
specific and potent inhibitor of NAADP (nicotinic acid adenine dinucleotide phosphate) signaling that suppresses Ca2+ oscillations


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library