Skip to Content
MilliporeSigma

SML2064

LBQ657

≥97% (HPLC), neprilysin inhibitor, powder

Synonym(s):

(2R,4S)-5-(Biphenyl-4-yl)-4-((3-carboxypropionyl)amino)-2-methylpentanoic acid, Desethyl sacubitril, Sacubitrilat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H25NO5
CAS Number:
Molecular Weight:
383.44
NACRES:
NA.77
UNSPSC Code:
12352200
MDL number:
eCl@ss:
32160703
Assay:
≥97% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

LBQ657, ≥97% (HPLC)

assay

≥97% (HPLC)

form

powder

optical activity

[α]/D -35 to -27°, c = 1 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(CCC(N[C@H](CC1=CC=C(C2=CC=CC=C2)C=C1)C[C@H](C(O)=O)C)=O)=O

InChI

1S/C22H25NO5/c1-15(22(27)28)13-19(23-20(24)11-12-21(25)26)14-16-7-9-18(10-8-16)17-5-3-2-4-6-17/h2-10,15,19H,11-14H2,1H3,(H,23,24)(H,25,26)(H,27,28)/t15-,19+/m1/s1

InChI key

DOBNVUFHFMVMDB-BEFAXECRSA-N

Biochem/physiol Actions

Inhibitor of neprilysin (neutral endopeptidase, NEP) formed by the action of esterases on its prodrug sacubitril (AHU377)
LBQ657 is the active metabolite of sacubitril. It is an inhibitor of the enkephalinase neprilysin (neutral endopeptidase, NEP) formed by the action of esterases on its prodrug sacubitril (AHU377), which is half of the heart failure combination drug Entresto, along with the angiotensin-receptor blocker Valsartan. LBQ657 prevents neprilysin′s degradation of atrial and brain natriuretic peptide, two blood pressure lowering peptides.


Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

STOT RE 1 - STOT SE 3

Target Organs

Bone,Kidney, Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library