Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2050

Sigma-Aldrich

AZ3451

≥98% (HPLC)

Synonym(s):

2-(6-Bromo-1,3-benzodioxol-5-yl)-N-(4-cyanophenyl)-1-[(1S)-1-cyclohexylethyl]-1H-benzimidazole-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H27BrN4O3
CAS Number:
Molecular Weight:
571.46
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N#CC1=CC=C(NC(C2=CC=C(N([C@H](C3CCCCC3)C)C(C4=CC(OCO5)=C5C=C4Br)=N6)C6=C2)=O)C=C1

Application

AZ3451 has been used as a protease-activated receptor 2 (PAR-2) antagonist to study the involvement of PAR-2 receptors in factor X activated (FXa)-induced upregulation of the cell adhesion molecules.

Biochem/physiol Actions

AZ3451 exhibits anti-inflammatory function in vivo. It can be used as a potential therapeutic for osteoarthritis (OA).
AZ3451 is a potent and selective allosteric antagonist of protease-activated receptor 2 (PAR2), a G protein-coupled receptor thought to be involved in inflammatory diseases, pain, allergic associated diseases (eosinophil migration, mast cells) and cancers. AZ3451 was found to bind to a remote allosteric site outside the helical bundle that is different from the site previously found on PAR1, and had an IC50 value of 23 nM. AZ3451 was able to completely inhibit ERK phosphorylation when added to 1321N1 cells overexpressing wild-type human PAR2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Schneckmann et al.
Basic research in cardiology, 118(1), 31-31 (2023-08-15)
Pharmacological inhibition of factor Xa by rivaroxaban has been shown to mediate cardioprotection and is frequently used in patients with, e.g., atrial fibrillation. Rivaroxaban's anti-inflammatory actions are well known, but the underlying mechanisms are still incompletely understood. To date, no
Robert K Y Cheng et al.
Nature, 545(7652), 112-115 (2017-04-27)
Protease-activated receptors (PARs) are a family of G-protein-coupled receptors (GPCRs) that are irreversibly activated by proteolytic cleavage of the N terminus, which unmasks a tethered peptide ligand that binds and activates the transmembrane receptor domain, eliciting a cellular cascade in
Xiaojian Huang et al.
Aging, 11(24), 12532-12545 (2019-12-17)
Osteoarthritis (OA) is a highly prevalent joint disorder blamed for pain and disability in older individuals. It's commonly accepted that inflammation, apoptosis, autophagy and cellular senescence participate in the progress of OA. Protease activated receptor 2 (PAR2), a member of
Cristina Almengló et al.
Biochemical pharmacology, 178, 114063-114063 (2020-06-04)
We aimed to study the effects of the new oral anticoagulant edoxaban, a factor X activated (FXa) inhibitor, on key endothelial functions that could contribute to cardiovascular benefit. Human umbilical endothelial cells (HUVEC) were obtained from donated umbilical cords and
Amanda J Kennedy et al.
Communications biology, 3(1), 782-782 (2020-12-19)
Protease-activated receptor-2 (PAR2) has been implicated in multiple pathophysiologies but drug discovery is challenging due to low small molecule tractability and a complex activation mechanism. Here we report the pharmacological profiling of a potent new agonist, suggested by molecular modelling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service