Skip to Content
MilliporeSigma

SML1932

A-1210477

≥98% (HPLC)

Synonym(s):

7-(1,3-Dimethyl-5-((4-(4-(methylsulfonyl)piperazin-1-yl)phenoxy)methyl)-1H-pyrazol-4-yl)-1-(2-morpholinoethyl)-3-(3-(naphthalen-1-yloxy)propyl)-1H-indole-2-carboxylic acid, 7-[5-[[4-[4-[(Dimethylamino)sulfonyl]-1-piperazinyl]phenoxy]methyl]-1,3-dimethyl-1H-pyrazol-4-yl]-1-[2-(4-morpholinyl)ethyl]-3-[3-(1-naphthalenyloxy)propyl]-1H-indole-2-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C46H55N7O7S
CAS Number:
Molecular Weight:
850.04
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(O)C(N1CCN2CCOCC2)=C(C3=C1C(C4=C(N(N=C4C)C)COC5=CC=C(C=C5)N6CCN(CC6)S(=O)(N(C)C)=O)=CC=C3)CCCOC7=C8C=CC=CC8=CC=C7.[H]Cl

InChI key

MRLDHNSBXWSDDL-UHFFFAOYSA-N

Biochem/physiol Actions

A BH3 mimetic that targets MCL-1 with high affinity and selectivity. Selectively inhibits cancer survival in a MCL-1-dependent manner.
A-1210477 is a BH3 mimetic that targets MCL-1 with high affinity (Ki = 0.43 nM) and selectivity (Ki >0.66 μM for BCL-2, BCL-XL, BCL-W, and A1), exhibiting little to no activity toward panels of 80 kinases (IC50 >8.5 μM) and 21 GPCRs (IC50 & EC50 >10 μM). A-1210477 selectively inhibits the survival of MCL-1-dependent (IC50 ~4 μM/multiple myeloma H929, 5.3 μM/NSCLC H2110, 7.2 μM/NSCLC H23), but not BCL-2-dependent RS4;11, cultures via apoptosis induction and potentiates BCL-2/BCL-XL inhibitor ABT-263 (Navitoclax) killing in a variety of cancer cell cultures where ABT-263 alone is ineffective. A-1210477 is also shown to cause cellular MCL-1 upregulation, most likely due to blocking MCL-1 interaction with BH3-only proteins such as NOXA and MULE known to mediate MCL-1 ubiquitylation for proteasomal degradation.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library