Skip to Content
MilliporeSigma

Skip To

SML1898

Iguratimod

≥98% (HPLC)

Synonym(s):

3-Formylamino-7-methylsulfonylamino-6-phenoxy-4H-1-benzopyran-4-one, IGU, N-(7-(Methylsulfonamido)-4-oxo-6-phenoxy-4H-chromen-3-yl)formamide, N-[3-(Formylamino)-4-oxo-6-phenoxy-4H-1-benzopyran-7-yl]methanesulfonamide, T-614

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H14N2O6S
CAS Number:
Molecular Weight:
374.37
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
eCl@ss:
32150313
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=S(NC1=CC(OC=C(NC([H])=O)C2=O)=C2C=C1OC3=CC=CC=C3)(C)=O

InChI

1S/C17H14N2O6S/c1-26(22,23)19-13-8-15-12(17(21)14(9-24-15)18-10-20)7-16(13)25-11-5-3-2-4-6-11/h2-10,19H,1H3,(H,18,20)

InChI key

ANMATWQYLIFGOK-UHFFFAOYSA-N

Biochem/physiol Actions

Iguratimod (T-614) is an orally active anti-inflammatory compound that inhibits NF-kappaB (NF-KB) p65 subunit (RelA) phosphorylation and nuclear translocation (30 μg/mL against 10 ng/mL TNF-α; human synovial cells) without affecting IKBα degradation.
Iguratimod (T-614) is an orally active anti-inflammatory compound that inhibits NF-kappaB p65 subunit (RelA) phosphorylation and nuclear translocation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1335
description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

room temp

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

H2O: 2 mg/mL, clear (warmed)

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters