Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1785

Sigma-Aldrich

Landiolol hydrochloride

≥98% (HPLC)

Synonym(s):

4-[(2S)-2-Hydroxy-3-[[2-[(4-morpholinylcarbonyl)amino]ethyl]amino]propoxy]-benzenepropanoic acid [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl ester hydrochloride, 4-[2-Hydroxy-3-[[2-[(4-morpholinylcarbonyl)amino]ethyl]amino]propoxy]-benzenepropanoic acid [S-(R*,R*)]-(2,2-dimethyl-1,3-dioxolan-4-yl)methyl ester hydrochloride, ONO 1101, ONO-1101

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H39N3O8 · HCl
CAS Number:
Molecular Weight:
546.05
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1(C)OC[C@@H](COC(CCC2=CC=C(OC[C@@H](O)CNCCNC(N3CCOCC3)=O)C=C2)=O)O1.[H]Cl

InChI

1S/C25H39N3O8.ClH/c1-25(2)35-18-22(36-25)17-34-23(30)8-5-19-3-6-21(7-4-19)33-16-20(29)15-26-9-10-27-24(31)28-11-13-32-14-12-28;/h3-4,6-7,20,22,26,29H,5,8-18H2,1-2H3,(H,27,31);1H/t20-,22+;/m0./s1

InChI key

DLPGJHSONYLBKP-IKGOIYPNSA-N

Biochem/physiol Actions

Landiolol is an ultra-short acting cardioselective β-adrenoceptor antagonist the exhibit β1-selectivity.
Landiolol is an ultra-short acting cardioselective β-adrenoceptor antagonist the exhibit β1-selectivity. New data shows that landiolol lacks pharmacochaperoning activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shahrooz Nasrollahi-Shirazi et al.
The Journal of pharmacology and experimental therapeutics, 359(1), 73-81 (2016-07-28)
Blockage of β1-adrenergic receptors is one of the most effective treatments in cardiovascular medicine. Esmolol was introduced some three decades ago as a short-acting β1-selective antagonist. Landiolol is a more recent addition. Here we compared the two compounds for their
Shigeki Kobayashi et al.
PloS one, 10(1), e0114314-e0114314 (2015-01-24)
The purpose of this study was to investigate whether adding a low-dose β1-blocker to milrinone improves cardiac function in failing cardiomyocytes and the underlying cardioprotective mechanism. The molecular mechanism underlying how the combination of low-dose β1-blocker and milrinone affects intracellular
Yoshiyasu Ogura et al.
Life sciences, 118(2), 347-356 (2014-05-23)
Endothelin (ET)-1 is the best known potent vasoconstrictor and has been implicated in pathogenesis of sepsis-associated acute kidney injury (AKI) in human or lipopolysaccharide (LPS)-induced AKI in animal models. We have previously shown that ET-1 is highly up-regulated in renal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service