Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1741

Sigma-Aldrich

TH1020

≥95% (HPLC)

Synonym(s):

4-((4-Benzyl-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl)thio)pyrido[3′,2′,4,5]-thieno[3,2-d]pyrimidine, 4-[[4-(Phenylmethyl)-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]thio]-pyrido[3′,2′:4,5]thieno[3,2-d]pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H15N7S2
CAS Number:
Molecular Weight:
453.54
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 3 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

C1(C2=CC=NC=C2)=NN=C(SC3=NC=NC4=C3SC5=C4C=CC=N5)N1CC6=CC=CC=C6

Application

TH1020 has been used to inhibit toll-like receptor 5 (TLR5) in porcine intestinal epithelial monolayer cells and mice.

Biochem/physiol Actions

TH1020 is a non-cytotoxic non-cytotoxic toll-like receptor 5 (TLR5)/flagellin complex inhibitor. TH1020 selectively prevents flagellin-induced TLR5 signaling without affecting ligands-induced activation of TLR2, TLR3, TLR4, TLR7 or TLR8.
TH1020 mediates the inhibition of tumor necrosis factor-α (TNF-α) based signaling, It disrupts tetrameric complex by binding the molecular interface between dimeric TLR5.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Luo et al.
Veterinary research, 50(1), 48-48 (2019-06-22)
Enterotoxigenic Escherichia coli (ETEC) are an important cause of post-weaning diarrhea (PWD) in piglets. The IL-17 cytokine family is well known to play important roles in the host defense against bacterial infections at the mucosa. Previously, we reported the potential
Lei Yan et al.
ChemMedChem, 11(8), 822-826 (2015-12-05)
Protein-protein interactions have been regarded as "undruggable" despite their importance in many biological processes. The complex formed between host toll-like receptor 5 (TLR5) and flagellin, a globular protein that is the main component of a bacterial flagellum, plays a vital role
Norimitsu Morioka et al.
Journal of neurochemistry, 150(6), 738-758 (2019-07-06)
Intrathecal treatment with recombinant high-mobility group box-1 (rHMGB1) in naïve mice leads to a persistent and significantly decreased hind paw withdrawal threshold to mechanical stimuli, suggesting that spinal HMGB1 evokes abnormal pain processing. By contrast, repeated intrathecal treatment with anti-HMGB1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service