Skip to Content
MilliporeSigma

SML1711

TNCA

≥98% (HPLC)

Synonym(s):

(3S)-2,3,4,9-Tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid, L-1,2,3,4-Tetrahydronorharman-3-carboxylic acid, Lycoperodine-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H12N2O2
CAS Number:
Molecular Weight:
216.24
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -57 to -67°, c = 1 (in MeOH:1N HCl (1:1))

color

white to beige

solubility

1.5 mg/mL, clear (Solvent: 0.1N HCl:DMSO (1:1); warmed)

storage temp.

−20°C

SMILES string

O=C(O)[C@H]1NCC2=C(C3=C(N2)C=CC=C3)C1

InChI

1S/C12H12N2O2/c15-12(16)10-5-8-7-3-1-2-4-9(7)14-11(8)6-13-10/h1-4,10,13-14H,5-6H2,(H,15,16)/t10-/m0/s1

InChI key

FSNCEEGOMTYXKY-JTQLQIEISA-N

Biochem/physiol Actions

TNCA is a tryptophan derivative that binds the parathyroid Ca2+-sensing receptor (CaSR) extracellular domain (ECD) hinge region between two globular subdomains and acts as a high-affinity CaSR ligand (CaSRL) with co-agonist activity.
TNCA is a tryptophan derivative that binds the parathyroid Ca2+-sensing receptor (CaSR) extracellular domain (ECD) hinge region between two globular subdomains and acts as a high-affinity CaSR ligand (CaSRL) with co-agonist activity. TNCA is shown to potentiate both Mg++- and Ca++-evoked cellular calcium response (0.1-0.5 mM), as well as Mg++-evoked cellular ERK1/2 phosphorylation in CaSR-expressing HEK293 cells (0.5 mM). AC-265347 (Cat. No. SML0129), on the other hand, is a calcimimetic that functions as a CaSR agonist.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library