Skip to Content
MilliporeSigma

SML1671

NCT-503 Inactive Control

≥98% (HPLC)

Synonym(s):

N-(4,6-Dimethylpyridin-2-yl)-4-(pyridin-4-yl)piperazine-1-carbothioamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H21N5S
CAS Number:
Molecular Weight:
327.45
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1=CC(NC(N2CCN(C3=CC=NC=C3)CC2)=S)=NC(C)=C1

InChI

1S/C17H21N5S/c1-13-11-14(2)19-16(12-13)20-17(23)22-9-7-21(8-10-22)15-3-5-18-6-4-15/h3-6,11-12H,7-10H2,1-2H3,(H,19,20,23)

InChI key

ITLPIAAGIQAGRA-UHFFFAOYSA-N

Application

NCT-503 Inactive Control has been used as a small molecule inhibitor of phosphoglycerate dehydrogenase in neuroblastoma cells.

Biochem/physiol Actions

NCT-503 Inactive Control is the inactive control probe for NCT-503, which is an inhibitor of phosphoglycerate dehydrogenase (PHGDH), the enzyme that catalyzes the first, rate-limiting step of glucose-derived serine synthesis. NCT-503 Inactive Control has an IC50 value >57 μM for PHGDH. NCT-503 has an IC50 value of 2.5 μM for PHGDH. NCT-503 Inactive Control is water soluble and has good stability, similar to NCT-503.
NCT-503 Inactive Control is the inactive control probe for NCT-503, which is an inhibitor of phosphoglycerate dehydrogenase (PHGDH).

Other Notes

The active PHGDH inhibitor, NCT-503, is available from Sigma. To learn more about and purchase NCT-503, click here.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library