Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1559

Sigma-Aldrich

Furamidine dihydrochloride

≥98% (HPLC)

Synonym(s):

2,5-Bis(4-amidinophenyl)furan dihydrochloride, 4,4′-(2,5-Furandiyl)bis-benzenecarboximidamide dihydrochloride, DB 75, DB75, NSC 305831, WR199385

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N4O · 2HCl
CAS Number:
Molecular Weight:
377.27
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 1 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

[Cl-].[Cl-].N[C+](N)c1ccc(cc1)c2[o]c(cc2)c3ccc(cc3)[C+](N)N

InChI

1S/C18H16N4O.2ClH/c19-17(20)13-5-1-11(2-6-13)15-9-10-16(23-15)12-3-7-14(8-4-12)18(21)22;;/h1-10H,(H3,19,20)(H3,21,22);2*1H

InChI key

VXNYQUQHOUERTR-UHFFFAOYSA-N

General description

Furamidine dihydrochloride is an analog of the aromatic drug pentamidine and a substrate of the organic cation transporter 1 (OCT1). A number of prodrugs of furamidine dihydrochloride are being synthesised and are in clinical trial stages.

Biochem/physiol Actions

Furamidine (DB75) binds to strings of AT base pair sequences in DNA′s minor groove. Furamidine was originally developed as an anti-parasitic compound for a variety of diseases including Chagas′ disease. Furamidine targets AT rich sequences in trypanosome kinetoplast minicircle DNA (kDNA), resulting in subsequent destruction of the kinetoplast and cell death. Furamidine has also been found to inihbit tyrosyl-DNA phosphodiesterase (Tdp1) and act as a selective protein arginine methyltransferase 1 (PRMT1) inhibitor with an IC50 of 9.4μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antiparasitic Compounds That Target DNA
Wilson
Biochimie, 90(7), 999-1014 (2008)
Xin Ming et al.
Drug metabolism and disposition: the biological fate of chemicals, 37(2), 424-430 (2008-10-31)
The antiparasitic activity of aromatic diamidine drugs, pentamidine and furamidine, depends on their entry into the pathogenic protozoa via membrane transporters. However, no such diamidine transporter has been identified in mammalian cells. The goal of this study is to investigate
Leilei Yan et al.
Journal of medicinal chemistry, 57(6), 2611-2622 (2014-02-26)
Protein arginine methylation is a posttranslational modification critical for a variety of biological processes. Misregulation of protein arginine methyltransferases (PRMTs) has been linked to many pathological conditions. Most current PRMT inhibitors display limited specificity and selectivity, indiscriminately targeting many methyltransferase
Yang Liu et al.
The journal of physical chemistry. B, 112(37), 11809-11818 (2008-08-23)
Given the increasing significance of diamidines as DNA-targeted therapeutics and biotechnology reagents, it is important to establish the variations in thermodynamic quantities that characterize the interactions of closely related compounds to different sequence AT binding sites. In this study, an
Smitha Antony et al.
Nucleic acids research, 35(13), 4474-4484 (2007-06-20)
By enzymatically hydrolyzing the terminal phosphodiester bond at the 3'-ends of DNA breaks, tyrosyl-DNA phosphodiesterase (Tdp1) repairs topoisomerase-DNA covalent complexes and processes the DNA ends for DNA repair. To identify novel Tdp1 inhibitors, we developed a high-throughput assay that uses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service