Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1543

Sigma-Aldrich

Apratastat

≥98% (HPLC)

Synonym(s):

(3S)-N-Hydroxy-4-[[4-[(4-hydroxy-2-butyn-1-yl)oxy]phenyl]sulfonyl]-2,2-dimethyl-3-thiomorpholinecarboxamide, (3S)-N-Hydroxy-4-[[4-[(4-hydroxy-2-butynyl)oxy]phenyl]sulfonyl]-2,2-dimethyl-3-thiomorpholinecarboxamide, TMI 005, TMI-005

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N2O6S2
CAS Number:
Molecular Weight:
414.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(N1CCSC(C)(C)[C@@H]1C(NO)=O)(C2=CC=C(OCC#CCO)C=C2)=O

InChI

1S/C17H22N2O6S2/c1-17(2)15(16(21)18-22)19(9-12-26-17)27(23,24)14-7-5-13(6-8-14)25-11-4-3-10-20/h5-8,15,20,22H,9-12H2,1-2H3,(H,18,21)/t15-/m0/s1

InChI key

MAVDNGWEBZTACC-HNNXBMFYSA-N

General description

Apratastat is one of the thiomorpholine sulfonamide hydroxymate selective inhibitors of ADAM17 (a disintegrin and metalloproteinase).

Biochem/physiol Actions

Apratastat (TMI-005) is an orally active, potent and selective dual inhibitor of disintegrin metalloenzyme 17 (ADAM17/ TACE) and matrix metalloprotease (MMP).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bridget M Ford et al.
American journal of physiology. Renal physiology, 305(3), F323-F332 (2013-05-17)
Matrix protein accumulation is a prominent feature of diabetic nephropathy that contributes to renal fibrosis and decline in renal function. The pathogenic mechanisms of matrix accumulation are incompletely characterized. We investigated if the matrix metalloprotease a disintegrin and metalloprotease1 7
Pharmacokinetic-pharmacodynamic modeling of apratastat: a population-based approach.
Shu C et al
Journal of Clinical Pharmacology, 51(4), 472-481 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service