Skip to Content
MilliporeSigma

SML1507

CRT0066101 hydrochloride

≥98% (HPLC)

Synonym(s):

2-[4-((R)-2-Amino-butylamino)-pyrimidin-2-yl]-4-(1-methyl-1Hpyrazol-4-yl)-phenol hydrochloride, 2-[4-[[(2R)-2-Aminobutyl]amino]-2-pyrimidinyl]-4-(1-methyl-1H-pyrazol-4-yl)-phenol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H22N6O · xHCl
CAS Number:
Molecular Weight:
338.41 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

Cl.[n]1(ncc(c1)c2cc(c(cc2)O)c3nc(ccn3)NC[C@H](N)CC)C

InChI

1S/C18H22N6O.ClH/c1-3-14(19)10-21-17-6-7-20-18(23-17)15-8-12(4-5-16(15)25)13-9-22-24(2)11-13;/h4-9,11,14,25H,3,10,19H2,1-2H3,(H,20,21,23);1H/t14-;/m1./s1

InChI key

NBKRZGHHBBHLOQ-PFEQFJNWSA-N

Biochem/physiol Actions

CRT0066101 is a very potent inhibitor of Protein Kinase D (PKD) isoforms with IC50 values for PKD-1, -2 and -3 = 1, 2.5 and 2 nM, respecitively. CRT0066101 has been shown to have anticancer activity. CRT0066101 potently inhibits proliferation of PANC-1 pancreatic cells, and blocks migration and invasion of U87MG glioblastoma cells. CRT0066101 has also been shown to decrease growth of primary tumors and metastasis of ER(-) breast cancers.
CRT0066101 is a very potent inhibitor of Protein Kinase D (PKD) isoforms.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library