Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1454

Sigma-Aldrich

Otilonium bromide

≥98% (HPLC)

Synonym(s):

N,N-Diethyl-N-methyl-2-[(4-{[2-(octyloxy)benzoyl]amino}benzoyl)oxy]ethanaminium bromide, N,N-Diethyl-N-methyl-2-[[4-[[2-(octyloxy)benzoyl]amino]benzoyl]oxy]-ethanaminium bromide, Octylonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H43BrN2O4
CAS Number:
Molecular Weight:
563.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

O=C(NC1=CC=C(C(OCC[N+](CC)(CC)C)=O)C=C1)C2=CC=CC=C2OCCCCCCCC.[Br-]

InChI

1S/C29H42N2O4.BrH/c1-5-8-9-10-11-14-22-34-27-16-13-12-15-26(27)28(32)30-25-19-17-24(18-20-25)29(33)35-23-21-31(4,6-2)7-3;/h12-13,15-20H,5-11,14,21-23H2,1-4H3;1H

InChI key

VWZPIJGXYWHBOW-UHFFFAOYSA-N

General description

Otilonium bromide is an antispasmodic. It contains a red color ACh (acetylcholine) like moiety bound to an aromatic system that ends with an octyl ether.

Biochem/physiol Actions

Otilonium Bromide is a muscarinic receptor antagonist (antimuscarinic). Otilonium Bromide is a quaternary ammonium salt that exerts spasmolytic action selective on the distal gastrointestinal tract. It is used world-wide for the treatment of irritable bowel syndrome (IBS).
Otilonium bromide has the capability to prevent L-type Ca2+ channels and colonic contractile activity. It is a tolerable drug as it is absorbed poorly and has very less side effects.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharma Chemica (2011)
Issues in Gastroenterology and Hepatology (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service