Skip to Content
MilliporeSigma

SML1433

AMD3465

≥98% (HPLC), CXCR4 antagonist, powder

Synonym(s):

AMD 3465, GENZ-644494, N-[[4-(1,4,8,11-Tetraazacyclotetradec-1-ylmethyl)phenyl]methyl]-2-pyridinemethanamine hexahydrobromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H38N6 · 6HBr
CAS Number:
Molecular Weight:
896.07
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

AMD3465, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

H2O: 25 mg/mL, clear

storage temp.

−20°C

SMILES string

N3(CCNCCCNCCNCCC3)Cc1ccc(cc1)CNCc2ncccc2

InChI

1S/C24H38N6/c1-2-13-29-24(5-1)20-28-19-22-6-8-23(9-7-22)21-30-17-4-12-26-15-14-25-10-3-11-27-16-18-30/h1-2,5-9,13,25-28H,3-4,10-12,14-21H2

InChI key

CWJJHESJXJQCJA-UHFFFAOYSA-N

Biochem/physiol Actions

AMD3465 is a highly selective chemokine receptor CXCR4 antagonist with antiviral and anticancer activity.
AMD3465 is a highly selective chemokine receptor CXCR4 antagonist with antiviral and anticancer activity. The chemokine receptor CXCR4 is expressed on a wide variety of leukocytes and is the predominant receptor for stromal cell-derived factor-1 (SDF1, CXCL12) in addition to acting as a co-receptor for HIV entry into cells. AMD3465 blocks surface binding of CXCL12 (SDF-1α). AMD3465 is an antagonist of SDF-1 ligand binding (K(i) of 41.7+/-1.2 nM). AMD3465 was found to halt the progression of an agressive childhood blood cancer, T-cell acute lymphoblastic leukemia (T-ALL), within two weeks of starting treatment in an animal model.
AMD3465 is also termed as{N-[1,4,8,11-tetraazacyclotetradecanyl-1,4-phenylenebis(methylene)]-2-(aminomethyl)pyridine}. It controls oncogenic signaling and invasiveness in vitro. It inhibits breast cancer growth and metastasis in vivo. As a chemokine receptor 4 (CXCR4) antagonist, AMD3465 is ten times more efficient than bicyclam AMD3100.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library