Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1415

Sigma-Aldrich

VE-821

≥98% (HPLC)

Synonym(s):

3-Amino-6-(4-(methylsulfonyl)phenyl)-N-phenylpyrazine-2-carboxamide, 3-Amino-6-[4-(methylsulfonyl)phenyl]-N-phenyl-2-pyrazinecarboxamide, VE821

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N4O3S
CAS Number:
Molecular Weight:
368.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥10 mg/mL, clear

storage temp.

−20°C

SMILES string

CS(C(C=C1)=CC=C1C2=NC(C(NC3=CC=CC=C3)=O)=C(N)N=C2)(=O)=O

InChI

1S/C18H16N4O3S/c1-26(24,25)14-9-7-12(8-10-14)15-11-20-17(19)16(22-15)18(23)21-13-5-3-2-4-6-13/h2-11H,1H3,(H2,19,20)(H,21,23)

InChI key

DUIHHZKTCSNTGM-UHFFFAOYSA-N

Application

VE-821 has been used as an inhibitor of ATM- and Rad3-related (ATR) protein in human cancer cells.

Biochem/physiol Actions

VE-821 is a potent ATP-competitive inhibitor of the DNA damage response (DDR) kinase Ataxia telangiectasia-mutated (ATM) and ATM- and Rad3-related (ATR) with a Ki of 13 nM. VE-821 has minimal cross-reactivity against the related PIKKs ATM, DNA-dependent protein kinase (DNA-PK), mTOR and PI3-kinase-γ (Ki of 16 μM, 2.2 μM, >1 μM and 3.9 μM, respectively) and against a large panel of unrelated protein kinases. VE-821 used alone caused death in a large fraction of cancer cell populations and also showed strong synergy with genotoxic agents. VE-821 increased sensitivity of cells to radiation and also sensitized cancer cells to a variety of chemotherapeutic agents.

Other Notes

VE-821 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the VE-821 probe summary on the Chemical Probes Portal website.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tingting Li et al.
Aging, 12(14), 14791-14807 (2020-07-21)
Protection of telomere 1 (POT1), the telomeric single-stranded DNA (ssDNA)-binding protein in the shelterin complex, has been implicated in the DNA damage response, tumorigenesis and aging. Telomere dysfunction induced by telomere deprotection could accelerate cellular senescence in primary human cells.
Human cancer cells utilize mitotic DNA synthesis to resist replication stress at telomeres regardless of their telomere maintenance mechanism
Ozgun Ozer, et al.
Oncotarget null
Aldo S Bader et al.
Nature communications, 14(1), 8419-8419 (2023-12-19)
DNA double-strand breaks (DSBs) are the most mutagenic form of DNA damage, and play a significant role in cancer biology, neurodegeneration and aging. However, studying DSB-induced mutagenesis is limited by our current approaches. Here, we describe iMUT-seq, a technique that
Yoichiro Harada et al.
eLife, 12 (2023-07-18)
Mannose has anticancer activity that inhibits cell proliferation and enhances the efficacy of chemotherapy. How mannose exerts its anticancer activity, however, remains poorly understood. Here, using genetically engineered human cancer cells that permit the precise control of mannose metabolic flux
Anastazja Poczta et al.
Scientific reports, 9(1), 14135-14135 (2019-10-03)
The present study investigated the effect of cladribine (CLA) and six of its derivatives containing a formamidine group at position 6 (CLA-FDM, CLA-FPAZ, CLA-FPIR, CLA-FPIP, CLA-FHEX, and CLA-FMOR) on acute promyelocytic, lymphoblastic, and acute monocytic leukemia cells. The role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service