Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1404

Sigma-Aldrich

A01 (Smurf1 inhibitor)

≥98% (HPLC)

Synonym(s):

[4-[[4-Chloro-3-(trifluoromethyl)phenyl]sulfonyl]-1-piperazinyl][4-(5-methyl-1H-pyrazol-1-yl)phenyl]methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20ClF3N4O3S
CAS Number:
Molecular Weight:
512.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=S(C1=CC=C(Cl)C(C(F)(F)F)=C1)(N2CCN(C(C3=CC=C(N4N=CC=C4C)C=C3)=O)CC2)=O

InChI

1S/C22H20ClF3N4O3S/c1-15-8-9-27-30(15)17-4-2-16(3-5-17)21(31)28-10-12-29(13-11-28)34(32,33)18-6-7-20(23)19(14-18)22(24,25)26/h2-9,14H,10-13H2,1H3

InChI key

QFYLTUDRXBNZFQ-UHFFFAOYSA-N

Biochem/physiol Actions

A01 is a high affinity selective inhibitor of E3 ubiquitin-protein ligase Smurf1 (Smad ubiquitination regulatory factor-1) that disturbs Smurf1-Smad1/5 interaction and blocks Smurf1 mediated Smad1/5 ubiquitination. A01 increases responsiveness to BMP-2 in myoblasts and osteoblasts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fanying Jiang et al.
Investigative ophthalmology & visual science, 65(2), 18-18 (2024-02-07)
TGF-β/BMP signaling pathway plays a significant role in fibrotic cataract. Smurf1, a ubiquitin protein ligase, regulates the TGF-β/BMP signaling pathway through the ubiquitin-proteasome system (UPS). This study aims to investigate the role of Smurf1 in the progression of fibrotic cataract
Haiyin Li et al.
Insect science, 29(5), 1251-1261 (2022-01-23)
Diapause, an important strategy used by insects to avoid adverse environments, is regulated by various cell signaling pathways. The results of our previous studies demonstrated that the transforming growth factor-β (TGF-β) signaling pathway regulated pupal diapause in Helicoverpa armigera, which
Julia Kaiser et al.
eNeuro, 7(5) (2020-09-06)
Transforming growth factor (TGF)β1 has repeatedly been associated with axonal regeneration and recovery after injury to the CNS. We found TGFβ1 upregulated in the stroke-denervated mouse spinal cord after ischemic injury to the motor cortex as early as 4 d

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service