Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1374

Sigma-Aldrich

Alamandine trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

(Ala¹)-Angiotensin I/II (1-7) trifluoroacetate salt, ARVYIHP trifluoroacetate salt, Angiotensin A (1-7) trifluoroacetate salt, H-Ala-Arg-Val-Tyr-Ile-His-Pro-OH trifluoroacetate salt, L-alanyl-L-arginyl-L-valyl-L-tyrosyl-L-isoleucyl-L-histidyl-L-Proline trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H62N12O9 · xC2HF3O2
CAS Number:
Molecular Weight:
855.00 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Alamandine is an endogenous heptapeptide that can be formed from angiotensin A by angiotensin converting enzyme-2 or directly from angiotensin-(1–7) by decarboxylation of its aspartate residue. Alamandine′s activity is similar to Ang-(1–7) rather than that of angiotensin II or angiotensin A. However, Alamandine acts at a different receptor, the Mas-related receptor, MrgD. Alamandine causes vasodilation, and has antifibrosis and antihypertensive activity.
Alamandine is an endogenous heptapeptide with similar activity to Angitensin (1-7), but acting at a different receptor, the Mas-related receptor, MrgD.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service