Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1323

Sigma-Aldrich

NU6140

≥98% (HPLC)

Synonym(s):

4-[[6-(Cyclohexylmethoxy)-9H-purin-2-yl]amino]-N,N-diethyl-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N6O2
CAS Number:
Molecular Weight:
422.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N(CC)CC)C1=CC=C(NC2=NC(NC=N3)=C3C(OCC4CCCCC4)=N2)C=C1

InChI

1S/C23H30N6O2/c1-3-29(4-2)22(30)17-10-12-18(13-11-17)26-23-27-20-19(24-15-25-20)21(28-23)31-14-16-8-6-5-7-9-16/h10-13,15-16H,3-9,14H2,1-2H3,(H2,24,25,26,27,28)

InChI key

XHEQSRJCJTWWAH-UHFFFAOYSA-N

Application

NU6140 has been used to study its effect on transcription factors NANOG, OCT4 (organic cation/carnitine transporter 4) and SOX2(SRY-box 2), involved in pluripotency of human embryonic stem (hES) cells and embryonal carcinoma-derived (hEC) cells.

Biochem/physiol Actions

NU6140 is a CDK2 inhibitor with 10-36-fold selectivity over other CDK family members. NU6140 induces apoptosis and cell cycle arrest in cancer cells in the G2-M phase, and potentiates the cytotoxic effects of paclitaxel.
NU6140 is known to downregulate the expression of survivin protein, which possesses antiapoptotic activity and is generally overexpressed in cancer cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment of the potential of CDK2 inhibitor NU6140 to influence the expression of pluripotency markers NANOG, OCT4, and SOX2 in 2102Ep and H9 cells.
Kallas A, et al.
International Journal of Cell Biology, 2014 (2014)
Marzia Pennati et al.
Molecular cancer therapeutics, 4(9), 1328-1337 (2005-09-20)
Cyclin-dependent kinases (CDK) play a crucial role in the control of the cell cycle. Aberrations in the control of cell cycle progression occur in the majority of human malignancies; hence, CDKs are promising targets for anticancer therapy. Here, we define

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service