Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1297

Sigma-Aldrich

SB269652

≥98% (HPLC)

Synonym(s):

N-[trans-4-[2-(7-Cyano-3,4-dihydro-2(1H)-isoquinolinyl)ethyl]cyclohexyl]-1H-indole-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30N4O
CAS Number:
Molecular Weight:
426.55
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL

storage temp.

2-8°C

SMILES string

[nH]1c2c(cc1C(=O)N[C@@H]3CC[C@H](CC3)CCN4CCc5c(cc(cc5)C#N)C4)cccc2

InChI

1S/C27H30N4O/c28-17-20-5-8-21-12-14-31(18-23(21)15-20)13-11-19-6-9-24(10-7-19)29-27(32)26-16-22-3-1-2-4-25(22)30-26/h1-5,8,15-16,19,24,30H,6-7,9-14,18H2,(H,29,32)/t19-,24-

InChI key

JGLGOAQPUQITLD-OGAOHHHESA-N

Biochem/physiol Actions

Allosteric modulator of D2 dopamine receptors
SB269652 is an allosteric modulator of D2 dopamine receptors. SB269652 binds to an allosteric site that is distinct from the orthosteric bound by typical D2 antagonists and is only revealed in D2 receptor dimers. SB269652 is bitopic, binding to an orthosteric site one one partner in the dimer and the allosteric site in the other. Engagement of this secondary pocket at the extracellular end of TM2 and TM7 is a new mechanism of modulating binding of a ligand to the orthosteric site of a second protomer by a G-protein coupled receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service