Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C20H19N7
CAS Number:
Molecular Weight:
357.41
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 10 mg/mL, clear
storage temp.
2-8°C
SMILES string
CN(N=C1)C=C1CCN2C=NC(C3=CN(C)N=C3)=C2C4=CC=C(C#N)C=C4
InChI
1S/C20H19N7/c1-25-12-16(10-23-25)7-8-27-14-22-19(18-11-24-26(2)13-18)20(27)17-5-3-15(9-21)4-6-17/h3-6,10-14H,7-8H2,1-2H3
InChI key
RRZVGDGTWNQAPW-UHFFFAOYSA-N
Biochem/physiol Actions
BAZ2-ICR is a chemical probe for BAZ2A/B bromodomains with >100-fold selectivity over other bromodomains, with the exception of CECR2 (15-fold selectivity). BAZ2A is an essential component of the nucleolar remodeling complex (NoRC), which mediates recruitment of histone modifyine enzymes and DNA methylase involved in the silencing of ribosomal RNA transcription by RNA polymerase I. BAZ2B is believed to be involved in regulating nucleosome mobilization along linear DNA. BAZ2-ICR binds to BAZ2A with a KD of 109 nM (ITC) and to BAZ2B with a KD of 170 nM (ITC). BAZ2-ICR also shows accelerated Fluorescence recovery after photobleaching (FRAP) recovery at 1 μM in the BAZ2A FRAP assay. For full characterization details, please visit the BAZ2-ICR probe summary on the Structural Genomics Consortium (SGC) website.
To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
BAZ2-ICR is a chemical probe for BAZ2A/B bromodomains.
Features and Benefits
BAZ2-ICR is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
Other Notes
BAZ2-ICR has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the BAZ2-ICR probe summary on the Chemical Probes Portal website.
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
