Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1270

Sigma-Aldrich

YZ9

≥98% (HPLC)

Synonym(s):

3-Carbethoxy-7-hydroxycoumarin, 7-hydroxy-2-oxo-2H-1-benzopyran-3-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O5
CAS Number:
Molecular Weight:
234.20
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

[o]1c2c(cc([c]1=O)C(=O)OCC)ccc(c2)O

InChI

1S/C12H10O5/c1-2-16-11(14)9-5-7-3-4-8(13)6-10(7)17-12(9)15/h3-6,13H,2H2,1H3

InChI key

IETDBZQIWIJQJG-UHFFFAOYSA-N

Biochem/physiol Actions

YZ9 is a 5, 6, 7, 8-tetrahydroxy-2-(4-hydroxyphenyl) chromen-4-one (N4A) inhibitor.
YZ9 is a potent inhibitor of PFKFB3 (6-phosphfructo-2-kinase/fructose-2,6-bisphosphatase) with an IC50 = 183 nM. PFKFB3 is over expressed in many cancers and catalyzes the production of fructose-2,6-bisphosphate, which can potentiate PFK1 activity, enhancing glycolysis in tumor cells. YZ9 dose dependently inhibits proliferation of HeLa cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minsuh Seo et al.
PloS one, 6(9), e24179-e24179 (2011-10-01)
Cancer cells adopt glycolysis as the major source of metabolic energy production for fast cell growth. The HIF-1-induced PFKFB3 plays a key role in this adaptation by elevating the concentration of Fru-2,6-BP, the most potent glycolysis stimulator. As this metabolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service