Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C6H14O8S2
CAS Number:
Molecular Weight:
278.30
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Product Name
Treosulfan, ≥98% (HPLC)
Quality Segment
assay
≥98% (HPLC)
form
powder
optical activity
[α]/D -4.9 to -5.9°, c = 1 in acetone
color
white to beige
solubility
H2O: 5 mg/mL, clear
storage temp.
−20°C
SMILES string
[S](=O)(=O)(OCC(O)C(O)CO[S](=O)(=O)C)C
InChI
1S/C6H14O8S2/c1-15(9,10)13-3-5(7)6(8)4-14-16(2,11)12/h5-8H,3-4H2,1-2H3
InChI key
YCPOZVAOBBQLRI-UHFFFAOYSA-N
Biochem/physiol Actions
Alkylating agent, anti-neoplastic agent
Treosulfan is a biofunctional alkylating agent.
Treosulfan is bifunctional alkylating agent.
Treosulfan/l-threitol-1,4-bis-methanesulfonate; dihydroxybusulfan; NSC 39069. It is a prodrug of a bifunctional alkylating cytotoxic agent. Treosulfan can be activated non-enzymatically to two active epoxides. It is used to treat ovarian cancer.
signalword
Danger
hcodes
Hazard Classifications
Carc. 1A - Muta. 1B - Repr. 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
