Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1247

Sigma-Aldrich

NAB2

≥98% (HPLC)

Synonym(s):

N-(2-Chlorobenzyl)-1-(2,5-dimethylphenyl)-1H-benzo[d]imidazole-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20ClN3O
CAS Number:
Molecular Weight:
389.88
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Clc1c(cccc1)CNC(=O)c2cc3nc[n](c3cc2)c4c(ccc(c4)C)C

InChI

1S/C23H20ClN3O/c1-15-7-8-16(2)22(11-15)27-14-26-20-12-17(9-10-21(20)27)23(28)25-13-18-5-3-4-6-19(18)24/h3-12,14H,13H2,1-2H3,(H,25,28)

InChI key

CZSLEMCYYGEGKP-UHFFFAOYSA-N

Biochem/physiol Actions

NAB2 is a cell permeable and selective modifier of α-synuclein toxicity diverse cell types and PD neurons. NAB2 restores Rsp5/Nedd4 E3 ligase-dependent endosomal and endoplasmic reticulum–to-Golgi vesicle trafficking.
NAB2 is a cell permeable and selective modifier of α-synuclein toxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service