Skip to Content

SML1241

Pyr6

≥98% (HPLC)

Synonym(s):

N-[4-[3,5-Bis(trifluoromethyl)-1H-pyrazol-1-yl]phenyl]-3-fluoro-4-pyridinecarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H9F7N4O
CAS Number:
Molecular Weight:
418.27
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=NC=C1F)NC2=CC=C(N3C(C(F)(F)F)=CC(C(F)(F)F)=N3)C=C2

InChI

1S/C17H9F7N4O/c18-12-8-25-6-5-11(12)15(29)26-9-1-3-10(4-2-9)28-14(17(22,23)24)7-13(27-28)16(19,20)21/h1-8H,(H,26,29)

InChI key

XZIQSOZOLJJMFN-UHFFFAOYSA-N

Biochem/physiol Actions

Pyr6 is a selective inhibitor of calcium release from the endoplasmic reticulum via STIM1/Orai1 mediated, store operated calcium entry (SOCE).
Pyr6 is a selective inhibitor of calcium release from the endoplasmic reticulum via STIM1/Orai1 mediated, store operated calcium entry (SOCE). Pyr6 blocks SOCE in thapsigargin treated, calcium depleted BRL-2H3 cells (IC50 = 0.49 μM), but has poor activity against carbachol-induced, TRPC3-mediated calcium entry in TRPC3-transfected HEK293 cells (IC50 = 18.46 μM).


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library