Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SML1228

Sigma-Aldrich

Cyclic-di-GMP sodium salt

≥98% (HPLC), powder, quorum sensing molecule

Synonym(s):

3′,5′-Cyclic diguanylic acid sodium salt, Bis(3′,5′)-cyclic diguanylic acid sodium salt, Cyclic Diguanosine-5′-monophosphate sodium salt, c-di-GMP sodium salt, c-diGMP sodium salt, cGpGp sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N10O14P2 · xNa+
CAS Number:
Molecular Weight:
690.41 (free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Cyclic-di-GMP sodium salt, ≥98% (HPLC)

description

contains 1μmol of powder (approx. 0.7mg)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

−20°C

SMILES string

O[C@@H]1[C@H](OP(OC[C@H]2O[C@@H](N3C(N=C(N)NC4=O)=C4N=C3)[C@H](O)[C@@H]2O5)(O)=O)[C@@H](COP5(O)=O)O[C@H]1N6C(N=C(N)NC7=O)=C7N=C6.C

InChI

1S/C20H24N10O14P2/c21-19-25-13-7(15(33)27-19)23-3-29(13)17-9(31)11-5(41-17)1-39-45(35,36)44-12-6(2-40-46(37,38)43-11)42-18(10(12)32)30-4-24-8-14(30)26-20(22)28-16(8)34/h3-6,9-12,17-18,31-32H,1-2H2,(H,35,36)(H,37,38)(H3,21,25,27,33)(H3,22,26,28,34)/t5-,6-,9-,10-,11-,12-,17-,18-/m1/s1

InChI key

PKFDLKSEZWEFGL-MHARETSRSA-N

Application

Cyclic-di-GMP sodium salt has been used as a quorum sensing molecule.

Biochem/physiol Actions

Cyclic-di-GMP is a naturally occurring small molecule that acts as a bacterial second messenger to regulate important signaling mechanisms in prokaryotes that control bacterial survival, adhesion, colonization, biofilm formation, and virulence factors production. During infections Cyclic-di-GMP is bound by host STING (stimulator of interferon genes) leading to expression of interferon genes, activating innate immune responses. c-di-GMP has been found in eukarotic systems acting not only in immune response, but as an inhibitor of the pacemaker I(f) current in the heart.

Preparation Note

Cyclic-di-GMP is soluble in water and aqueous buffers.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Angel De la Cruz-Villegas et al.
Journal of microbiology and biotechnology, 30(6), 811-821 (2020-04-03)
Mycobacterium tuberculosis produces mycolic acids which are relevant for persistence, recalcitrance to antibiotics and defiance to host immunity. c-di-GMP is a second messenger involved in transition from planktonic cells to biofilms, whose levels are controlled by diguanylate cyclases (DGC) and
An Aphrodisiac Produced By Vibrio fischeri Stimulates Mating In The Closest Living Relatives Of Animals.
Woznica A, et al.
bioRxiv, 139022-139022 (2017)
Lei Zhang et al.
Nature communications, 11(1), 5371-5371 (2020-10-25)
Autoinducer-2 (AI-2) is a quorum sensing signal that mediates communication within and between many bacterial species. However, its known receptors (LuxP and LsrB families) are not found in all the bacteria capable of responding to this signaling molecule. Here, we
Andrea Annibal et al.
Analytical and bioanalytical chemistry, 413(26), 6457-6468 (2021-09-04)
Cyclic dinucleotides (CDNs) are key secondary messenger molecules produced by cyclic dinucleotide synthases that trigger various cellular signaling cascades from bacteria to vertebrates. In mammals, cyclic GMP-AMP synthase (cGAS) has been shown to bind to intracellular DNA and catalyze the
Emily N Chin et al.
Science (New York, N.Y.), 369(6506), 993-999 (2020-08-21)
Stimulator of interferon genes (STING) links innate immunity to biological processes ranging from antitumor immunity to microbiome homeostasis. Mechanistic understanding of the anticancer potential for STING receptor activation is currently limited by metabolic instability of the natural cyclic dinucleotide (CDN)

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service