Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C35H36N4O5
CAS Number:
Molecular Weight:
592.68
UNSPSC Code:
12161501
PubChem Substance ID:
MDL number:
Assay:
≥90% (HPLC)
Form:
powder
Storage condition:
protect from light
assay
≥90% (HPLC)
form
powder
storage condition
protect from light
color
blue, to dark brown to black
solubility
DMSO: 0.5 mg/mL, clear (warmed)
storage temp.
−20°C
SMILES string
O=C([C@@H]1C(OC)=O)C(C(N/2)=C1C([C@@H](CCC(O)=O)[C@@H]/3C)=NC3=C/4)=C(C)C2=C\C5=N/C(C(C)=C5CC)=C\C6=C(C=C)C(C)=C4N6
InChI
1S/C35H36N4O5/c1-8-19-15(3)22-12-24-17(5)21(10-11-28(40)41)32(38-24)30-31(35(43)44-7)34(42)29-18(6)25(39-33(29)30)14-27-20(9-2)16(4)23(37-27)13-26(19)36-22/h8,12-14,17,21,31,36,39H,1,9-11H2,2-7H3,(H,40,41)/b22-12-,23-13-,24-12-,25-14-,26-13-,27-14-,32-30-/t17-,21-,31+/m0/s1
InChI key
NSFSLUUZQIAOOX-QEWKCGBTSA-N
Application
Pheophorbide-a has been used as a photosensitizer:
- for the photodynamic therapy (PDT) in oral cancer cell lines
- to test its effect on inducing oxidative stress and death in L. amazonensis
- to test its effect on the viability of HeLa cells
Biochem/physiol Actions
Pheophorbide-a, due to its photosensitizing effect, may serve as a promising bioactive compound to treat leishmaniasis. It is a metabolite of chlorophyll and possesses immunostimulation functionality. Pheophorbide-a is a photosensitizer for the photodynamic therapy. It is an ATP Binding Cassette Subfamily G Member 2 (ABCG2) transporter specific substrate. Overexpression of ATP-binding cassette (ABC) transporters in cancer cells is associated with the multidrug resistance phenotype.
Other Notes
light sensitive
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.