Skip to Content
MilliporeSigma

SML1149

BMS-189453

≥98% (HPLC)

Synonym(s):

(E)-4-[2-(5,6-Dihydro-5,5-dimethyl-8-phenyl-2-naphthalenyl)ethenyl]-benzoic acid, BMS-453, BMS453

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H24O2
CAS Number:
Molecular Weight:
380.48
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

OC(C(C=C1)=CC=C1/C=C/C(C=C2)=CC3=C2C(C)(C)CC=C3C4=CC=CC=C4)=O

InChI

1S/C27H24O2/c1-27(2)17-16-23(21-6-4-3-5-7-21)24-18-20(12-15-25(24)27)9-8-19-10-13-22(14-11-19)26(28)29/h3-16,18H,17H2,1-2H3,(H,28,29)/b9-8+

InChI key

VUODRPPTYLBGFM-CMDGGOBGSA-N

Biochem/physiol Actions

BMS-189453 is a potent RARβ agonist that acts as an antagonist against RARα and RARγ. BMS-189453 induces RARβ reporter gene expression at sub nanomolar levels, and is 30 fold more potent than all-trans retinoic acid for inducing TGFβ activity in normal breast cells. The compound BMS-189453 does not transactivate RARα or γ transcriptional activity, but binding to those family members induces a strong transrepression of phorbol ester-induced AP-1 activity (IC50 = 0.1 nM in HeLa and MCSF-7). BMS-189453 significantly increases the efficiency of cardiac differentiation of hESCs.
BMS-189453 is a potent RARβ agonist.
BMS-189453 specifically has sufficient bioavailability in rats and monkeys. BMS-189453 only binds to α, β, and γ retinoid receptors but its activation is unknown. BMS-189453 is found to inhibit the action of collagenase-3 (MMP-13) which catalyses cartilage matrix degradation. Thus, it serves to treat rheumatoid arthritis.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library