Skip to Content
MilliporeSigma

SML1060

Celecoxib-NBD

≥98% (HPLC)

Synonym(s):

N-(2-((7-Nitrobenzo[c]-[1,2,5]oxadiazol-4-yl)amino)ethyl)-4-(5-(p-tolyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl)benzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H20F3N7O5S
Molecular Weight:
587.53
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

yellow to orange

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Celecoxib-NBD is a flourescent COX2-specific inhibitor.
Celecoxib-NBD is a flourescent COX2-specific inhibitor. The molecule binds to, and inhibits COX2 with an IC50 of 0.19 μM, and a selectivity index of 443 for COX2 vs. COX1. Celecoxib-NBD fluorescently labels COX2-expressing HCA-7 colon cells, but not COX2-negative HCT116 cells.


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library