Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1014

Sigma-Aldrich

CYM-5520

≥98% (HPLC)

Synonym(s):

1-[2-(1-Benzyl-2,5-dimethyl-1H-pyrrol-3-yl)-2-oxo-ethyl]-6-oxo-1,6-dihydro-pyridine-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19N3O2
CAS Number:
Molecular Weight:
345.39
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Application

CYM-5520 has been used as a sphingosine 1-phosphate receptor 2 (S1pr2) agonist in pancreatic adenocarcinoma cell line (CFPAC-1).

Biochem/physiol Actions

CYM-5520 is a specific, noncompetitive allosteric agonist of sphingosine 1-phosphate receptor 2 (S1PR2; EC50 = 1.0 μM). The compound CYM-5520 does not show any agonsit activity towards S1PR1, S1PR3, S1PR4 or S1PR5.
CYM-5520 is a specific, noncompetitive allosteric agonist of sphingosine 1-phosphate receptor 2.
CYM-5520 mediates phosphorylation of transcription factor p65. It upregulates trabecular number, thickness as well as tibial bone volume density in osteoblasts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filamin A expression negatively regulates sphingosine-1-phosphate-induced NF-kappaB activation in melanoma cells by inhibition of Akt signaling
Campos LS, et al.
Molecular and Cellular Biology, 36(2), 320-329 (2016)
Sphingosine-1-phosphate/S1PR2-mediated signaling triggers Smad1/5/8 phosphorylation and thereby induces Runx2 expression in osteoblasts
Higashi K, et al.
Bone, 93, 1-11 (2016)
p120 catenin suppresses basal epithelial cell extrusion in invasive pancreatic neoplasia
Hendley AM, et al.
Cancer Research, 76(11), 3351-3363 (2016)

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service