Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1005

Sigma-Aldrich

NSC 617145

≥98% (HPLC)

Synonym(s):

(2,3-Propandiyl-bis(2,3-dichloro)maleimide, 1,1′-(2,2-Dimethyl-1,3-propanediyl)bis[3,4-dichloro-1H-Pyrrole-2,5-dione, NSC-617145, SID 487795

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10Cl4N2O4
CAS Number:
Molecular Weight:
400.04
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 3 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C13H10Cl4N2O4/c1-13(2,3-18-9(20)5(14)6(15)10(18)21)4-19-11(22)7(16)8(17)12(19)23/h3-4H2,1-2H3

InChI key

PCOXPBOKDABARQ-UHFFFAOYSA-N

Biochem/physiol Actions

NSC 617145 has an IC50 value of 230nM.
NSC 617145 is a potent and cell-permeable inhibitor of Werner syndrome helicase (WRN) that causes accumulation of double-strand breaks (DSB) and chromosomal abnormalities in cancer cells.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

DNA repair in cancer therapy, 257-257 (2011)

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service