Skip to Content
MilliporeSigma

SML0971

Ki 16425

≥98% (HPLC)

Synonym(s):

3-[[[4-[4-[[[1-(2-Chlorophenyl)ethoxy]carbonyl]amino]-3-methyl-5-isoxazolyl]phenyl]methyl]thio]-propanoic acid, Ki16425

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H23ClN2O5S
CAS Number:
Molecular Weight:
474.96
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

S(CCC(=O)O)Cc1ccc(cc1)c2[o]nc(c2NC(=O)OC(C)c3c(cccc3)Cl)C

InChI

1S/C23H23ClN2O5S/c1-14-21(25-23(29)30-15(2)18-5-3-4-6-19(18)24)22(31-26-14)17-9-7-16(8-10-17)13-32-12-11-20(27)28/h3-10,15H,11-13H2,1-2H3,(H,25,29)(H,27,28)

InChI key

LLIFMNUXGDHTRO-UHFFFAOYSA-N

Application

Ki 16425 has been used as a chemical inhibitor to study the regulation of LPA (lysophosphatidic acid) on LPAR(LPA receptor) subtypes.

Biochem/physiol Actions

Ki16452 is a potent antagonist of the lysophosphatidic acid receptors LPA1 and LPA3, with greater than 30-fold selectivity for LPA1 over LPA2. The Ki values for LPA1 and LPA3 are 250 nM and 360 nM, respectively.
Ki16452 is a potent antagonist of the lysophosphatidic acid receptors LPA1 and LPA3.
Ki 16425 possesses a short-lived inhibitory activity. It has been studied that Ki 16425 is effective in the inhibition of neuropathic pain‐like behaviors.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library