Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0950

Sigma-Aldrich

Meclizine dihydrochloride

≥97% (HPLC)

Synonym(s):

1-(p-Chloro-α-phenylbenzyl)-4-(m-methylbenzyl)-piperazine dihydrochloride, 1-[(4-Chlorophenyl)phenylmethyl]4-[?(3-methylphenyl)methyl]piperazine dihydrochloride, NSC 28728

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27ClN2 · 2HCl
CAS Number:
Molecular Weight:
463.87
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].Clc1ccc(cc1)C(N3CCN(CC3)Cc4cc(ccc4)C)c2ccccc2.[H+].[H+]

InChI

1S/C25H27ClN2.2ClH/c1-20-6-5-7-21(18-20)19-27-14-16-28(17-15-27)25(22-8-3-2-4-9-22)23-10-12-24(26)13-11-23;;/h2-13,18,25H,14-17,19H2,1H3;2*1H

InChI key

VCTHNOIYJIXQLV-UHFFFAOYSA-N

Biochem/physiol Actions

Meclizine has the ability to increase glycolysis and decrease cellular ATP levels in media that has glucose and galactose. It guards the kidney against ischemia-reperfusion injury. Meclizine is considered as an anti-nausea and anti-dizziness drug, was identified in a ′nutrient-sensitized′ chemical screen. In human, it acts as an agonist of pregnane X receptor (PXR). It enhances the expression of CYP3A4 (cytochrome P450 3A4) mRNA and reduces CYP3A-catalyzed testosterone 6β-hydroxylation in primary cultures of human hepatocytes.
Meclizine is a potent histamine H1 receptor antagonist that is widely used as antiemetic.
Meclizine is a potent histamine H1 receptor antagonist that is widely used as antiemetic. Recently, meclizine was identified as an inhibitor of hepstin. Also, meclizine is a potent inhibitor of mitochondrial respiration through inhibition of cytosolic CTP:phosphoethanolamine cytidylyltransferase (Pcyt2).

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meclizine, a pregnane X receptor agonist, is a direct inhibitor and mechanism-based inactivator of human cytochrome P450 3A.
Foo WYB, et al.
Biochemical Pharmacology, 97(3), 320-330 (2015)
Meclizine Preconditioning Protects the Kidney Against Ischemia-Reperfusion Injury.
Kishi S, et al.
EBioMedicine, 2(9), 1090-1101 (2015)

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service