Skip to Content
MilliporeSigma

SML0933

OL-135

≥98% (HPLC)

Synonym(s):

1-Oxo-1-[5-(2-pyridyl)oxazol-2-yl]-7-phenylheptane, 7-Phenyl-1-[5-(2-pyridinyl)-2-oxazolyl]-1-heptanone, OL 135

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H22N2O2
CAS Number:
Molecular Weight:
334.41
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

n1c([o]c(c1)c3ncccc3)C(=O)CCCCCCc2ccccc2

InChI

1S/C21H22N2O2/c24-19(14-7-2-1-4-10-17-11-5-3-6-12-17)21-23-16-20(25-21)18-13-8-9-15-22-18/h3,5-6,8-9,11-13,15-16H,1-2,4,7,10,14H2

InChI key

ILOIOIGZFHGSMS-UHFFFAOYSA-N

Biochem/physiol Actions

OL-135 is a CNS penetrant, highly potent and selective reversible inhibitor of FAAH devoid of CB1, CB2 and μ and δ opioid receptors activities that increases the analgesic and hypothermic activity of anandamide.
OL-135 is a CNS penetrant, highly potent and selective reversible inhibitor of FAAH.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library