Skip to Content
MilliporeSigma

SML0819

Bisdionin C

≥98% (HPLC)

Synonym(s):

1,1′-(1,3-Propanediyl)bis[3,7-dihydro-3,7-dimethyl-1H-purine-2,6-dione, 1,1′-Trimethylenedi-theobromine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H20N8O4
CAS Number:
Molecular Weight:
400.39
NACRES:
NA.77
UNSPSC Code:
51111800
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Bisdionin C is a cell-permeable, competitive acidic mammalian chitinase (AMCase) and chitotriosidase (CHIT1) inhibitor.
Bisdionin C is a cell-permeable, competitive acidic mammalian chitinase (AMCase) and chitotriosidase (CHIT1) inhibitor. Bisdionin C also potently inhibits bacterial AfChiB1 chitinase.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library