Skip to Content
MilliporeSigma

SML0814

ESI-09

≥98% (HPLC)

Synonym(s):

α-[2-(3-Chlorophenyl)hydrazinylidene]-5-(1,1-dimethylethyl)-b-oxo-3-isoxazolepropanenitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H15ClN4O2
CAS Number:
Molecular Weight:
330.77
UNSPSC Code:
41106305
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

, faint yellow to dark orange

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1cc(ccc1)N\N=C(\C(=O)c2n[o]c(c2)C(C)(C)C)/C#N

InChI

1S/C16H15ClN4O2/c1-16(2,3)14-8-12(21-23-14)15(22)13(9-18)20-19-11-6-4-5-10(17)7-11/h4-8,19H,1-3H3/b20-13+

InChI key

DXEATJQGQHDURZ-DEDYPNTBSA-N

Application

ESI-09 has been used as an antagonist for exchange factor directly activated by cAMP (EPAC).

Biochem/physiol Actions

ESI-09 is a potent, specific inhibitor of EPAC (exchange protein directly activated by cAMP).
ESI-09 is a potent, specific inhibitor of EPAC (exchange protein directly activated by cAMP). ESI-09 inhibits EPAC1 and EPAC2 with IC50 values of 3.2 and 1.4 μM, respectively, with no activity against PKA at 25 μM. In pancreatic cell lines, the compound blocks phosphorylation of Akt and insulin secretion. ESI-09 inhibits migration of pancreatic cancer cell lines.


Still not finding the right product?

Explore all of our products under ESI-09


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library