Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C32H43N5O2
CAS Number:
Molecular Weight:
529.72
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12171501
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 5 mg/mL, clear
storage temp.
2-8°C
SMILES string
O=C(N1CCC(N2CCCC2)CC1)C3=CC(NC4=CC=CC=C4)=C(C(N5CCC(N6CCCC6)CC5)=O)C=C3
InChI
1S/C32H43N5O2/c38-31(36-20-12-27(13-21-36)34-16-4-5-17-34)25-10-11-29(30(24-25)33-26-8-2-1-3-9-26)32(39)37-22-14-28(15-23-37)35-18-6-7-19-35/h1-3,8-11,24,27-28,33H,4-7,12-23H2
InChI key
PQOOIERVZAXHBP-UHFFFAOYSA-N
Biochem/physiol Actions
UNC1215 is a potent, selective antagonist of L3MBTL3 with cellular activity.
UNC1215 is a potent, selective antagonist of L3MBTL3 with cellular activity. Lethal (3) malignant brain tumor-like protein 3 (L3MBTL3) is a putative polycomb group (PcG) protein with gene-repressing activity, a member of the MBT family of methyllysine binders. Methyl-lysine binding proteins are a family of proteins that are important epigenetic regulators that “read” the post-translational methylation state of histone lysine residues and modulate protein-protein interactions that regulate gene expression. UNC1215 has an IC50 of 20 nM and > 100-fold selectivity over 13 HMTs and selected representatives of kinases, ion channels, 7TMs, and other epigenetic proteins. UNC1215 showed a significant decrease in FRAP recovery (Fluorescence Recovery After Photobleaching) time below 1 microM in cells using GFP-L3MBTL3. For full characterization details, please visit the UNC1215 probe summary on the Structural Genomics Consortium (SGC) website.
To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
Features and Benefits
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
UNC1215 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC
Other Notes
UNC1215 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the UNC1215 probe summary on the Chemical Probes Portal website.
1 of 1
This Item | |||
|---|---|---|---|
| description ≥98% (HPLC) | description - | description - | description ≥98% (HPLC) |
| form powder | form - | form - | form powder |
| assay ≥98% (HPLC) | assay - | assay - | assay ≥98% (HPLC) |
| Quality Level 100 | Quality Level - | Quality Level - | Quality Level 100 |
| storage temp. 2-8°C | storage temp. - | storage temp. - | storage temp. 2-8°C |
| solubility DMSO: 5 mg/mL, clear | solubility - | solubility - | solubility DMSO: 15 mg/mL, clear |
| color white to beige | color - | color - | color white to beige |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



