Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0640

Sigma-Aldrich

2-Methylserotonin maleate

≥95% (HPLC)

Synonym(s):

2-Me-5-HT maleate, 2-Methyl-5-HT maleate, 2-Methyl-5-hydroxytryptamine maleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O · C4H4O4
CAS Number:
Molecular Weight:
306.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to brown

solubility

H2O: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC2=C(NC(C)=C2CCN)C=C1.OC(/C=C\C(O)=O)=O

InChI

1S/C11H14N2O.C4H4O4/c1-7-9(4-5-12)10-6-8(14)2-3-11(10)13-7;5-3(6)1-2-4(7)8/h2-3,6,13-14H,4-5,12H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

KFEAUMZKRNJEDU-BTJKTKAUSA-N

Biochem/physiol Actions

2-Methylserotonin is a selective full agonist at the 5-HT3 receptor.
2-Methylserotonin maleate, a serotonin type 3 (5-HT3) agonist is used as a training drug in drug discrimination studies. Use of 2-methylserotonin in evolving rats shows to reflect the ontogeny of the spinal 5-HT3 receptor system.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5-HT3 agonist 2-methylserotonin as a training drug in drug discrimination studies
Glennon R A et al.
Pharmacology, Biochemistry, and Behavior, 41(2), 361-364 (1992)
Antinociceptive effects of intrathecally administered 2-methylserotonin in developing rats
Giordano J
Dev. Brain Res., 98(1), 142-144 (1997)

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service