Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0608

Sigma-Aldrich

A-412997 dihydrochloride

≥98% (HPLC)

Synonym(s):

N-(3-Methylphenyl)-4-(2-pyridinyl)-1-piperidineacetamide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N3O · 2HCl
CAS Number:
Molecular Weight:
382.33
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

Cl.Cl.N2(CCC(CC2)c3ncccc3)CC(=O)Nc1cc(ccc1)C

InChI

1S/C19H23N3O.2ClH/c1-15-5-4-6-17(13-15)21-19(23)14-22-11-8-16(9-12-22)18-7-2-3-10-20-18;;/h2-7,10,13,16H,8-9,11-12,14H2,1H3,(H,21,23);2*1H

InChI key

KIGNSSHMCNBHQX-UHFFFAOYSA-N

Biochem/physiol Actions

A-412997 is a selective D4 agonist with Ki values of 12 and 7.9 nM for the rat and human receptors, and no affinity for other dopamine receptors at concentrations up to 1 mM. A-412997 improves short term memory and cognitive properties in rodent models.
A-412997 is a selective D4 agonist.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service