Skip to Content
MilliporeSigma

SML0528

BIM5078

≥98% (HPLC)

Synonym(s):

1-(2′-Chloro-5′-nitrobenzenesulfonyl)-2-methylbenzimidazole, 1-[(2-Chloro-5-nitrophenyl)sulfonyl]-2-methyl-1H-benzimidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H10ClN3O4S
CAS Number:
Molecular Weight:
351.76
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1=NC2=CC=CC=C2N1S(C3=C(Cl)C=CC([N+]([O-])=O)=C3)(=O)=O

InChI

1S/C14H10ClN3O4S/c1-9-16-12-4-2-3-5-13(12)17(9)23(21,22)14-8-10(18(19)20)6-7-11(14)15/h2-8H,1H3

InChI key

DLUVMRPVZILBOE-UHFFFAOYSA-N

Biochem/physiol Actions

BIM5078 is a potent Hepatocyte nuclear factor 4a (HNF4a) antagonist that directly binds to ligand-binding pocket and modulates the expression of known HNF4a target genes. BIM5078 also inhibits insulin gene expression by disruption of E47 and PDX-1 binding to insulin promoter. BIM5078 is selectively cytotoxic to transformed cells.
BIM5078 is a potent Hepatocyte nuclear factor 4α antagonist.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library