Skip to Content
MilliporeSigma

Skip To

SML0501

SB-590885

≥98% (HPLC)

Synonym(s):

5-(2-(4-(2-Dimethylaminoethoxy)phenyl)-5-(4-pyridinyl)-1H-imidazol-4-yl)-2,3-dihydro-1H-inden-1-one oxime

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C27H27N5O2
CAS Number:
Molecular Weight:
453.54
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL (clear solution, warmed)

storage temp.

2-8°C

SMILES string

CN(C)CCOc1ccc(cc1)-c2nc(-c3ccc4c(CC\C4=N\O)c3)c([nH]2)-c5ccncc5

InChI

1S/C27H27N5O2/c1-32(2)15-16-34-22-7-3-19(4-8-22)27-29-25(18-11-13-28-14-12-18)26(30-27)21-5-9-23-20(17-21)6-10-24(23)31-33/h3-5,7-9,11-14,17,33H,6,10,15-16H2,1-2H3,(H,29,30)/b31-24-

InChI key

MLSAQOINCGAULQ-QLTSDVKISA-N

Application

SB-590885 has been used as a component of a mixture to maintain naive human pluripotent stem cells (PSCs).It has also been used as B-Raf (BRAF) inhibitor to study its effects on lung adenocarcinoma cell lines.

Biochem/physiol Actions

SB-590885 is a potent and selective ATP competitive inhibitor of B-Raf kinase with Kd = 300 pM for B-Raf, and >1000-fold selectivity over a panel of 22 commonly studied cellular kinases
SB-590885 is a potent and selective B-Raf kinase inhibitor.

Features and Benefits

This compound is featured on the MAPKKKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2825S7067SML2212
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL (clear solution, warmed)

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

faintly yellow, to beige

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters