Skip to Content
MilliporeSigma

SML0473

Noladin ether

≥98% (HPLC)

Synonym(s):

2-Arachidonylglyceryl ether, 2-[(5Z,8Z,11Z,14Z)-5,8,11,14-Eicosatetraen-1-yloxy]-1,3-propanediol; 2-AG ether, HU 310

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H40O3
CAS Number:
Molecular Weight:
364.56
UNSPSC Code:
12352200
EC Number:
200-578-6
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
solution
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solution

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

concentration

10 mg/mL in ethanol (1% w/v)

color

colorless to light yellow

shipped in

dry ice

storage temp.

−70°C

SMILES string

O(C(CO)CO)CCCC\C=C/C\C=C/C\C=C/C\C=C/CCCCC

InChI

1S/C23H40O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-26-23(21-24)22-25/h6-7,9-10,12-13,15-16,23-25H,2-5,8,11,14,17-22H2,1H3/b7-6-,10-9-,13-12-,16-15-

InChI key

CUJUUWXZAQHCNC-DOFZRALJSA-N

Biochem/physiol Actions

Noladin ether (2-Arachidonyl glycerol ether) is a CB1 agonist.
Noladin ether (2-Arachidonyl glycerol ether) is a stable analog of the endogenous cannabanoid 2-Arachidonylglycerol (2-AG) with selectivity for CB1 receptors. Noladin ether is a CB1 agonist. The Ki values for CB1 vs CB2 receptors are 21.2 nM and 3 mM, respectively. Noladin ether induces sedation and has modest antinociceptive activity in vivo, and reduces inter ocular pressure after administration in the eye.

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

57.2 °F - closed cup

flash_point_c

14.0 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library