Skip to Content

SML0466

LCS-1

≥98% (HPLC)

Synonym(s):

4,5-Dichloro-2-(3-methylphenyl)-3(2H)-pyridazinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H8Cl2N2O
CAS Number:
Molecular Weight:
255.10
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solutikon, warmed)

storage temp.

2-8°C

SMILES string

O=C1C(Cl)=C(Cl)C=NN1C2=CC=CC(C)=C2

InChI

1S/C11H8Cl2N2O/c1-7-3-2-4-8(5-7)15-11(16)10(13)9(12)6-14-15/h2-6H,1H3

InChI key

SYUPLLHVMCLXEM-UHFFFAOYSA-N

Biochem/physiol Actions

LCS-1 is an inhibitor of superoxide dismutase (SOD1).
LCS-1 is an inhibitor of superoxide dismutase (SOD1). LCS-1 inhibits the proliferation of lung adendocarcinoma cell lines.


Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library