Skip to Content
MilliporeSigma

SML0448

URB937

≥95% (HPLC)

Synonym(s):

Cyclohexylcarbamic acid 3′–carbamoyl–6–hydroxybiphenyl–3–yl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H22N2O4
CAS Number:
Molecular Weight:
354.40
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder (1:1 ratio of product to ethanol)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder (1:1 ratio of product to ethanol)

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

N(C3CCCCC3)C(=O)Oc1cc(c(cc1)O)c2cc(ccc2)C(=O)N

InChI

1S/C20H22N2O4/c21-19(24)14-6-4-5-13(11-14)17-12-16(9-10-18(17)23)26-20(25)22-15-7-2-1-3-8-15/h4-6,9-12,15,23H,1-3,7-8H2,(H2,21,24)(H,22,25)

InChI key

CMEQHOXCIGFZNJ-UHFFFAOYSA-N

Biochem/physiol Actions

URB937 is a fatty acid amide hydrolase (FAAH) inhibitor that is actively excluded from the CNS by the membrane transporter ABCG2. The ED50 for inhibition of FAAH activity in the brain is 200 times higher than inhibition of renal FAAH activity. In vivo administration of URB937 elevates anandamide levels in peripheral tissues, and attenuates acetic acid-induced writhing, hyperalgesia in a sciatic nerve ligation model, and pain and edema in carrageenan injected paws.
URB937 is a fatty acid amide hydrolase (FAAH) inhibitor.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library