Skip to Content
MilliporeSigma

SML0174

GPR35 Agonist, Compound 10

≥98% (HPLC)

Synonym(s):

2-[4-[(2,4-Dioxo-5-thiazolidinylidene)methyl]phenoxy]-acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H9NO5S
CAS Number:
Molecular Weight:
279.27
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality level:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥5 mg/mL at ~60 °C

storage temp.

2-8°C

SMILES string

OC(=O)COc1ccc(cc1)\C=C2/SC(=O)NC2=O

InChI

1S/C12H9NO5S/c14-10(15)6-18-8-3-1-7(2-4-8)5-9-11(16)13-12(17)19-9/h1-5H,6H2,(H,14,15)(H,13,16,17)/b9-5-

InChI key

WXMCOLGPDOYHNK-UITAMQMPSA-N

Application

GPR35 Agonist, Compound 10, may be used in GPR35-mediated cell signaling studies.

Biochem/physiol Actions

GPR35 Agonist
GPR35 agonist Compound 10 is a GPR35 agonist the binds with higher affinity than other known agonists, such as kynurenic acid and zaprinast. Compound 10 was more potent than zaniprast (pEC50 = 5.3 vs 4.18) for recombinantly expressed human GPR35, and unlike zaprinast, is equipotent for human and rat GPR35


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ida Marie Modvig et al.
American journal of physiology. Endocrinology and metabolism, 320(5), E874-E885 (2021-03-02)
The aim of this study was to explore individual amino acid-stimulated GLP-1 responses and the underlying stimulatory mechanisms, as well as to identify the amino acid-sensing receptors involved in amino acid-stimulated GLP-1 release. Experiments were primarily based on isolated perfused



Global Trade Item Number

SKUGTIN
SML0174-5MG04061832982304
SML0174-25MG04061832260723