Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML0096

Sigma-Aldrich

Cinnabarinic Acid

≥98% (HPLC)

Synonym(s):

2-amino-3-oxo-3H-phenoxazine-1,9-dicarboxylic acid, Cinnabaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8N2O6
CAS Number:
Molecular Weight:
300.22
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

red to very dark red

solubility

DMSO: ≥4 mg/mL

storage temp.

2-8°C

SMILES string

NC1=C(C(O)=O)C2=Nc3c(OC2=CC1=O)cccc3C(O)=O

InChI

1S/C14H8N2O6/c15-10-6(17)4-8-12(9(10)14(20)21)16-11-5(13(18)19)2-1-3-7(11)22-8/h1-4H,15H2,(H,18,19)(H,20,21)

InChI key

FSBKJYLVDRVPTK-UHFFFAOYSA-N

Application

Cinnabarinic acid may be used in studies of the functions of components of the kynurenine metabolic pathway. It may be used to study it role as a metabotropic glutamate receptor (mGlu4R-specific) agonist.

Biochem/physiol Actions

Caspase Inducer; mGlu4R agonist
Cinnabarinic acid (CA) connects between initiation of the kynurenine pathway and immune tolerance that is used to prevent neuroinflammation.
Cinnabarinic acid is a kynurenine pathway metabolite of tryptophan, produced by the oxidation of 3-Hydroxyanthranilic acid. Cinnabarinic acid leads to loss of mitochondrial respiration and apoptosis, and has also been shown to be an mGlu4R-specific agonist.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

C Eggert
Microbiological research, 152(3), 315-318 (1997-11-14)
Concentrated culture fluid of the wood-rotting basidiomycete Pycnoporus cinnabarinus showed biological activity against a variety of bacterial strains. The maximal inhibitory effect was obtained for Gram-positive bacteria of the genus Streptococcus. In general, inhibition was higher for Gram-positive than Gram-negative
J A Dykens et al.
Biochemical pharmacology, 36(2), 211-217 (1987-01-15)
The oxidative reactivities of four tryptophan metabolites in the kynurenine pathway were examined as a potential mechanism for their reported neurotoxicities and carcinogenicities. Neither quinolinic acid, a neurotoxin, nor its monocarboxylic analogue, picolinic acid, auto-oxidized over a wide pH range.
H Iwahashi
Journal of chromatography. B, Biomedical sciences and applications, 736(1-2), 237-245 (2000-02-17)
3-Hydroxyanthranilic acid (3-HAA)-derived oxidation products were analyzed using high-performance liquid chromatography with an electrochemical reactor and diode array detection and high-performance liquid chromatography with an electrochemical reactor and UV detection coupled with mass spectrometry. In addition to 3-HAA dimers such
M K Manthey et al.
Biochimica et biophysica acta, 1034(2), 207-212 (1990-05-16)
The autoxidation of the tryptophan metabolite, 3-hydroxyanthranilic acid, at pH 7 gives rise to a p-quinone dimer and cinnabarinic acid. A novel dimer formed by radical-radical coupling of 3-hydroxyanthranilic acid is also produced. Labelling studies have shown that the C-2
H Ogawa et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 364(8), 1059-1066 (1983-08-01)
Cinnabarinic acid was formed from 3-hydroxyanthranilic acid during incubation with a soluble fraction from Malpighian tubules of the silkworm, Bombyx mori, in the presence of manganese ion. The enzyme having this activity was purified to homogeneity by ammonium sulfate fractionation

Articles

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service