Skip to Content
MilliporeSigma

SML0091

L-Dopa ethyl ester

≥98% (HPLC), Dopaminergic acent, powder

Synonym(s):

3,4-Dihydroxy-L-phenylalanine ethyl ester, 3-Hydroxy-L-tyrosine ethyl ester, Etilevodopa, L-Dopa ethyl ester, L-Dopa-Et, Levodopa ethyl ester, O-Ethyl-DOPA, TV-1203

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H15NO4
CAS Number:
Molecular Weight:
225.24
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Dopa ethyl ester, ≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +15 to +19°, c = 1 in methanol ([α]/D)

color

white to tan

solubility

H2O: ≥12 mg/mL at warmed to 60 °C

storage temp.

2-8°C

SMILES string

CCOC(=O)[C@@H](N)Cc1ccc(O)c(O)c1

InChI

1S/C11H15NO4/c1-2-16-11(15)8(12)5-7-3-4-9(13)10(14)6-7/h3-4,6,8,13-14H,2,5,12H2,1H3/t8-/m0/s1

InChI key

NULMGOSOSZBEQL-QMMMGPOBSA-N

Gene Information

human ... DRD3(1814)

Application

L-Dopa ethyl ester has been used to study the protective effects of piperlongumine (PLG) on rotenone-induced Parkinson disease (PD).

Biochem/physiol Actions

L-Dopa ethyl ester is a prodrug of levodopa that has greater gastric solubility. The drug is rapidly hydrolyzed to levodopa by nonspecific esterases in the gastrointestinal tract. L-Dopa ethyl ester (etilevodopa)-carbidopa treatment was well tolerated but did not demonstrate better efficacy compared with standard levodopa-carbidopa treatment. L-Dopa ethyl ester should have a significant brain penetration following an administration by injection.
L-Dopa ethyl ester is prodrug of levodopa that has greater gastric solubility; antiparkinsonian agent
L-Dopa ethyl ester (L-Dopa) is a dopamine replacement drug. It is used as a potent therapeutic for Parkinson disease (PD). L-Dopa has some long-term side effects, such as the deterioration of motor function.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library