Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB01052

Sigma-Aldrich

Aridanin

≥90% (LC/MS-ELSD)

Synonym(s):

3-({2-Deoxy-2-[(1-hydroxyethylidene)amino]hexopyranosyl}oxy)olean-12-en-28-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H61NO8
CAS Number:
Molecular Weight:
659.89
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

659.89

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C38H61NO8/c1-21(41)39-28-30(43)29(42)24(20-40)46-31(28)47-27-12-13-35(6)25(34(27,4)5)11-14-37(8)26(35)10-9-22-23-19-33(2,3)15-17-38(23,32(44)45)18-16-36(22,37)7/h9,23-31,40,42-43H,10-20H2,1-8H3,(H,39,41)(H,44,45)/t23-,24+,25-,26+,27-,28+,29+,30+,31-,35-,36+,37+,38-/m0/s1

InChI key

VRFWJSCLROXBBW-FUHHSGJXSA-N

General description

Aridanin is a triterpenoid saponin belongs to group of pentacyclic triterpenes. It is a natural product commonly isolated from Tetrapleura tetraptera, Albizia subdimidiata and Adenanthera pavanina. Existing research indicates that this metabolite, derived from plant source, exhibits various biological properties, including antibacterial, antioxidant, anti-inflammatory, and anticancer activities.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Aridanin exibits strong molluscicidal properties against the schistosomiasis-transmitting snails Biomphalaria glabrata.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adetunji J Aladesanmi
African journal of traditional, complementary, and alternative medicines : AJTCAM, 4(1), 23-36 (2006-01-01)
Tetrapleura tetraptera (Schumach. And Thonn) Taub, Mimosaceae, commonly known as Aridan (fruit), A single stemmed, robust, perennial tree of about 30 m. It has a grey/brown, smooth/rough bark with glabrous yound branchlets. The flower is yellow/pink and racemes white the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service