Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB01049

Sigma-Aldrich

4′-O-Methylabyssinone V

≥90% (LC/MS-ELSD)

Synonym(s):

4-Methylabyssinone V, 5,7-Dihydroxy-4′-methoxy-3′,5′-diprenylflavanone, Abyssinone V 4′-methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30O5
CAS Number:
Molecular Weight:
422.51
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

422.51

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

General description

4′-O-Methylabyssinone V, a prenylated flavonone, is a natural product commonly found in plants of Erythrina species. It has shown antioxidant, anti-inflammatory, antimicrobial, and anticancer properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical and biochemical research.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service