Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB01048

Sigma-Aldrich

Uzarin

≥90% (LC/MS-ELSD)

Synonym(s):

(3beta,5alpha)-3-((6-O-beta-D -glucopyranosyl-beta-D -glucopyranosyl)oxy)-14-hydroxycard-20(22)-enolide, Uzarigenin 3-β-sophoroside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H54O14
CAS Number:
Molecular Weight:
698.79
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

698.79

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1CC(=CC1=O)[C@@H]2[C@@]3([C@@]([C@H]4[C@@H]([C@@]5([C@H](C[C@H](CC5)O[C@@H]6O[C@@H]([C@H]([C@@H]([C@H]6O[C@@H]7O[C@@H]([C@H]([C@@H]([C@H]7O)O)O)CO)O)O)CO)CC4)C)CC3)(CC2)O)C

InChI

1S/C35H54O14/c1-33-8-5-18(46-32-30(28(42)26(40)23(14-37)48-32)49-31-29(43)27(41)25(39)22(13-36)47-31)12-17(33)3-4-21-20(33)6-9-34(2)19(7-10-35(21,34)44)16-11-24(38)45-15-16/h11,17-23,25-32,36-37,39-44H,3-10,12-15H2,1-2H3/t17-,18-,19+,20-,21+,22+,23+,25+,26+,27-,28-,29+,30+,31-,32+,33-,34+,35-/m0/s1

InChI key

RIPMDUQTRRLJTE-QHYHXNCWSA-N

General description

Uzarin, also known as Uzarigenin, is a cardenolide glycoside commonly found in plants such as Xysmalobium undulatum, Nerium oleander, Asclepias fruticosa, and Calotropis procera. Existing research suggests that this plant-derived metabolite exerts multiple biological properties, including antidiarrheal, antiviral and anticancer.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

According to the existing research, Uzarin plays potential role as an inhibitor of the SARS-CoV-2 receptor binding domain and SARS-CoV-2 RNA-dependent polymerase, as indicated by molecular docking and dynamics studies.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service